Assay Details
| Gene Symbol: AT4G11120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117182.5 | NP_192850.2 | 6 - 7 | 1052 | 61 | |
| GenBank mRNA | BT015741.1 | - | 6 - 7 | 973 | 61 | |
| BT020174.1 | - | 5 - 6 | 930 | 61 | ||
More Information
Set Membership: |
|

AT4G11120
Genomic Map