Assay Details
| Gene Symbol: TUF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117185.5 | NP_192853.1 | 5 - 6 | 860 | 63 | |
| GenBank mRNA | AY065119.1 | - | 5 - 6 | 785 | 63 | |
| AY081632.1 | - | 5 - 6 | 656 | 63 | ||
More Information
Set Membership: |
|

TUF
Genomic Map