Assay Details
| Gene Symbol: AT4G11300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117201.4 | NP_192869.1 | 1 | 1026 | 65 | |
| GenBank mRNA | AK221630.1 | - | 1 | 878 | 65 | |
| BT025658.1 | - | 1 | 797 | 65 | ||
More Information
Set Membership: |
|

AT4G11300
Genomic Map