Assay Details
| Gene Symbol: GPX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117229.4 | NP_192897.2 | 2 - 3 | 396 | 113 | |
| GenBank mRNA | AY039863.1 | - | 2 - 3 | 345 | 113 | |
| AY077655.1 | - | 2 - 3 | 320 | 113 | ||
More Information
Set Membership: |
|

GPX6
Genomic Map