Assay Details
| Gene Symbol: GPX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117229.4 | NP_192897.2 | 5 - 6 | 742 | 76 | |
| GenBank mRNA | AY039863.1 | - | 5 - 6 | 691 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPX6| Gene Symbol: GPX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117229.4 | NP_192897.2 | 5 - 6 | 742 | 76 | |
| GenBank mRNA | AY039863.1 | - | 5 - 6 | 691 | 76 | |
Set Membership: |
|