Assay Details
| Gene Symbol: HAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117241.2 | NP_192909.2 | 2 - 3 | 370 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HAP2| Gene Symbol: HAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117241.2 | NP_192909.2 | 2 - 3 | 370 | 92 | |
Set Membership: |
|