Assay Details
| Gene Symbol: HMGS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117251.3 | NP_192919.1 | 1 - 2 | 267 | 88 | |
| GenBank mRNA | U79160.1 | - | 1 - 2 | 181 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HMGS| Gene Symbol: HMGS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117251.3 | NP_192919.1 | 1 - 2 | 267 | 88 | |
| GenBank mRNA | U79160.1 | - | 1 - 2 | 181 | 88 | |
Set Membership: |
|