Assay Details
| Gene Symbol: AGL14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340739.1 | NP_001319907.1 | 3 - 4 | 575 | 77 | |
| NM_001340740.1 | NP_001329521.1 | 3 - 4 | 575 | 77 | ||
| NM_001340741.1 | NP_001329520.1 | 3 - 4 | 575 | 77 | ||
| NM_117258.5 | NP_192925.1 | 3 - 4 | 382 | 77 | ||
| GenBank mRNA | AK230464.1 | - | 3 - 4 | 383 | 77 | |
More Information
Set Membership: |
|

AGL14
Genomic Map