Assay Details
| Gene Symbol: CCS52A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117262.3 | NP_192929.2 | 3 - 4 | 868 | 95 | |
| GenBank mRNA | AY063875.1 | - | 3 - 4 | 823 | 95 | |
| AY091235.1 | - | 3 - 4 | 698 | 95 | ||
More Information
Set Membership: |
|

CCS52A2
Genomic Map