Assay Details
| Gene Symbol: CCS52A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117262.3 | NP_192929.2 | 6 - 7 | 1412 | 86 | |
| GenBank mRNA | AY063875.1 | - | 6 - 7 | 1367 | 86 | |
| AY091235.1 | - | 6 - 7 | 1242 | 86 | ||
More Information
Set Membership: |
|

CCS52A2
Genomic Map