Assay Details
| Gene Symbol: CCS52A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117262.3 | NP_192929.2 | 8 - 9 | 1542 | 90 | |
| GenBank mRNA | AY063875.1 | - | 8 - 9 | 1497 | 90 | |
| AY091235.1 | - | 8 - 9 | 1372 | 90 | ||
More Information
Set Membership: |
|

CCS52A2
Genomic Map