Assay Details
| Gene Symbol: PGRL1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203773.1 | NP_001190702.1 | 7 - 8 | 749 | 159 | |
| NM_117266.4 | NP_192933.2 | 6 - 7 | 786 | 159 | ||
| GenBank mRNA | AK117721.1 | - | 6 - 7 | 656 | 159 | |
| BT005305.1 | - | 6 - 7 | 607 | 159 | ||
More Information
Set Membership: |
|

PGRL1B
Genomic Map