Assay Details
Gene Symbol: NIP1;2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118008.4 | NP_193626.1 | 4 - 5 | 834 | 81 | |
GenBank mRNA | AJ250668.1 | - | 4 - 5 | 727 | 81 | |
AY072380.1 | - | 4 - 5 | 690 | 81 | ||
BT000096.1 | - | 4 - 5 | 667 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
