Assay Details
| Gene Symbol: AT4G27640 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118900.4 | NP_194494.2 | 13 - 14 | 2261 | 109 | |
| GenBank mRNA | AY062727.1 | - | 13 - 14 | 2242 | 109 | |
| BT010736.1 | - | 13 - 14 | 2155 | 109 | ||
More Information
Set Membership: |
|

AT4G27640
Genomic Map