Assay Details
| Gene Symbol: AT4G27640 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118900.4 | NP_194494.2 | 14 - 15 | 2379 | 84 | |
| GenBank mRNA | AY062727.1 | - | 14 - 15 | 2360 | 84 | |
| BT010736.1 | - | 14 - 15 | 2273 | 84 | ||
More Information
Set Membership: |
|

AT4G27640
Genomic Map