Assay Details
| Gene Symbol: AT4G27640 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118900.4 | NP_194494.2 | 3 - 4 | 652 | 68 | |
| GenBank mRNA | AY062727.1 | - | 3 - 4 | 633 | 68 | |
| BT010736.1 | - | 3 - 4 | 546 | 68 | ||
More Information
Set Membership: |
|

AT4G27640
Genomic Map