Assay Details
| Gene Symbol: ACBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341882.1 | NP_001328474.1 | 2 - 3 | 815 | 62 | |
| NM_118916.3 | NP_194507.1 | 2 - 3 | 815 | 62 | ||
| GenBank mRNA | AF178947.1 | - | 2 - 3 | 780 | 62 | |
| AF320561.1 | - | 2 - 3 | 805 | 62 | ||
| AY088510.1 | - | 2 - 3 | 815 | 62 | ||
| AY090256.1 | - | 2 - 3 | 802 | 62 | ||
| AY122892.1 | - | 2 - 3 | 590 | 62 | ||
More Information
Set Membership: |
|

ACBP2
Genomic Map