Assay Details
| Gene Symbol: ACBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118916.3 | NP_194507.1 | 4 - 5 | 1006 | 58 | |
| GenBank mRNA | AF178947.1 | - | 4 - 5 | 971 | 58 | |
| AF320561.1 | - | 4 - 5 | 996 | 58 | ||
| AY088510.1 | - | 4 - 5 | 1006 | 58 | ||
| AY090256.1 | - | 4 - 5 | 993 | 58 | ||
| AY122892.1 | - | 4 - 5 | 781 | 58 | ||
More Information
Set Membership: |
|

ACBP2
Genomic Map