Assay Details
| Gene Symbol: AT4G27880 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118926.3 | NP_194517.1 | 2 - 3 | 1072 | 83 | |
| GenBank mRNA | BT033078.1 | - | 2 - 3 | 616 | 83 | |
More Information
Set Membership: |
|

AT4G27880
Genomic Map