Assay Details
| Gene Symbol: AT5G20090 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085137.1 | NP_001078606.1 | 4 - 5 | 406 | 116 | |
| NM_001343637.1 | NP_001330067.1 | 4 - 5 | 569 | 116 | ||
| NM_122016.3 | NP_197509.1 | 4 - 5 | 452 | 116 | ||
| GenBank mRNA | AY050938.1 | - | 4 - 5 | 368 | 116 | |
| AY085758.1 | - | 4 - 5 | 376 | 116 | ||
More Information
Set Membership: |
|

AT5G20090
Genomic Map