Assay Details
| Gene Symbol: SUS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036838.2 | NP_001031915.1 | 2 - 3 | 382 | 135 | |
| NM_001343687.1 | NP_001332603.1 | 2 - 3 | 182 | 135 | ||
| NM_122090.4 | NP_197583.1 | 1 - 2 | 781 | 135 | ||
| GenBank mRNA | AK316826.1 | - | 1 - 2 | 188 | 135 | |
More Information
Set Membership: |
|

SUS1
Genomic Map