Assay Details
| Gene Symbol: SUS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036838.2 | NP_001031915.1 | 10 - 11 | 1752 | 98 | |
| NM_001343687.1 | NP_001332603.1 | 10 - 11 | 1552 | 98 | ||
| NM_122090.4 | NP_197583.1 | 9 - 10 | 2151 | 98 | ||
| GenBank mRNA | AK316826.1 | - | 9 - 10 | 1558 | 98 | |
More Information
Set Membership: |
|

SUS1
Genomic Map