Assay Details
| Gene Symbol: AT5G20890 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122097.3 | NP_197589.1 | 8 - 9 | 899 | 69 | |
| GenBank mRNA | AY054210.1 | - | 8 - 9 | 806 | 69 | |
| AY062651.1 | - | 8 - 9 | 811 | 69 | ||
| AY085104.1 | - | 8 - 9 | 806 | 69 | ||
| BT002090.1 | - | 7 - 8 | 636 | 69 | ||
More Information
Set Membership: |
|

AT5G20890
Genomic Map