Assay Details
| Gene Symbol: JAZ12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122098.5 | NP_197590.1 | 2 - 3 | 413 | 105 | |
| GenBank mRNA | AF360184.1 | - | 2 - 3 | 328 | 105 | |
| AY051013.1 | - | 2 - 3 | 234 | 105 | ||
More Information
Set Membership: |
|

JAZ12
Genomic Map