Assay Details
| Gene Symbol: AIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122099.4 | NP_197591.1 | 1 - 2 | 358 | 74 | |
| GenBank mRNA | AY081329.1 | - | 1 - 2 | 297 | 74 | |
| BT000238.1 | - | 1 - 2 | 165 | 74 | ||
| DQ059130.1 | - | 1 - 2 | 165 | 74 | ||
More Information
Set Membership: |
|

AIP2
Genomic Map