Assay Details
| Gene Symbol: AIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122099.4 | NP_197591.1 | 3 - 4 | 769 | 96 | |
| GenBank mRNA | AY081329.1 | - | 3 - 4 | 708 | 96 | |
| BT000238.1 | - | 3 - 4 | 576 | 96 | ||
| DQ059130.1 | - | 3 - 4 | 576 | 96 | ||
More Information
Set Membership: |
|

AIP2
Genomic Map