Assay Details
| Gene Symbol: TSL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122101.2 | NP_568405.1 | 13 - 14 | 2094 | 118 | |
| GenBank mRNA | L23985.1 | - | 13 - 14 | 2054 | 118 | |
More Information
Set Membership: |
|

TSL
Genomic Map