Assay Details
| Gene Symbol: TSL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122101.2 | NP_568405.1 | 1 - 2 | 596 | 97 | |
| GenBank mRNA | L23985.1 | - | 1 - 2 | 556 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TSL| Gene Symbol: TSL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122101.2 | NP_568405.1 | 1 - 2 | 596 | 97 | |
| GenBank mRNA | L23985.1 | - | 1 - 2 | 556 | 97 | |
Set Membership: |
|