Assay Details
| Gene Symbol: B73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343692.1 | NP_001332397.1 | 8 - 9 | 2073 | 92 | |
| NM_001343693.1 | NP_001332398.1 | 8 - 9 | 1316 | 92 | ||
| NM_122108.4 | NP_197599.1 | 9 - 10 | 1576 | 92 | ||
More Information
Set Membership: |
|

B73
Genomic Map