Assay Details
| Gene Symbol: LARP1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203425.1 | NP_001190354.1 | 5 - 6 | 1248 | 143 | |
| NM_001203426.1 | NP_001190355.1 | 5 - 6 | 1248 | 143 | ||
| NM_122123.3 | NP_568409.1 | 5 - 6 | 1245 | 143 | ||
| GenBank mRNA | AY052365.1 | - | 5 - 6 | 1166 | 143 | |
| AY139801.1 | - | 5 - 6 | 1030 | 143 | ||
More Information
Set Membership: |
|

LARP1a
Genomic Map