Assay Details
| Gene Symbol: AT5G22400 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122145.5 | NP_197632.1 | 1 - 2 | 706 | 66 | |
| GenBank mRNA | AK229223.1 | - | 1 - 2 | 520 | 66 | |
| BT026360.1 | - | 1 - 2 | 467 | 66 | ||
More Information
Set Membership: |
|

AT5G22400
Genomic Map