Assay Details
| Gene Symbol: AT5G22470 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343735.1 | NP_001318618.1 | 5 - 6 | 819 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT5G22470| Gene Symbol: AT5G22470 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343735.1 | NP_001318618.1 | 5 - 6 | 819 | 62 | |
Set Membership: |
|