Assay Details
| Gene Symbol: F8H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122200.4 | NP_197685.2 | 1 - 2 | 845 | 84 | |
| GenBank mRNA | BT011629.1 | - | 1 - 2 | 533 | 84 | |
| BT014962.1 | - | 1 - 2 | 310 | 84 | ||
| KJ138671.1 | - | 1 - 2 | 310 | 84 | ||
More Information
Set Membership: |
|

F8H
Genomic Map