Assay Details
| Gene Symbol: F8H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343782.1 | NP_001330046.1 | 1 - 2 | 1022 | 125 | |
| NM_122200.4 | NP_197685.2 | 2 - 3 | 1211 | 125 | ||
| GenBank mRNA | BT011629.1 | - | 2 - 3 | 899 | 125 | |
| BT014962.1 | - | 2 - 3 | 676 | 125 | ||
| KJ138671.1 | - | 2 - 3 | 676 | 125 | ||
More Information
Set Membership: |
|

F8H
Genomic Map