Assay Details
| Gene Symbol: MYB37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122206.4 | NP_197691.1 | 2 - 3 | 367 | 68 | |
| GenBank mRNA | AK175507.1 | - | 2 - 3 | 367 | 68 | |
| AY519628.1 | - | 2 - 3 | 268 | 68 | ||
| DQ446976.1 | - | 2 - 3 | 268 | 68 | ||
More Information
Set Membership: |
|

MYB37
Genomic Map