Assay Details
| Gene Symbol: AT5G23550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122262.3 | NP_197746.1 | 5 - 6 | 649 | 73 | |
| GenBank mRNA | AK117540.1 | - | 5 - 6 | 600 | 73 | |
| BT005168.1 | - | 5 - 6 | 451 | 73 | ||
More Information
Set Membership: |
|

AT5G23550
Genomic Map