Assay Details
| Gene Symbol: AT5G23600 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122266.2 | NP_197750.1 | 1 | 563 | 106 | |
| GenBank mRNA | BT029507.1 | - | 1 | 256 | 106 | |
| Gene Symbol: AT5G23590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122265.5 | NP_197749.2 | 1 | 627 | 106 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_122265.5 transcripts.
Set Membership: |
|

AT5G23600
Genomic Map