Assay Details
| Gene Symbol: LCB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036855.1 | NP_001031932.1 | 6 - 7 | 879 | 82 | |
| NM_122272.3 | NP_197756.1 | 6 - 7 | 895 | 82 | ||
| GenBank mRNA | AY050829.1 | - | 6 - 7 | 842 | 82 | |
| AY133827.1 | - | 6 - 7 | 647 | 82 | ||
More Information
Set Membership: |
|

LCB2
Genomic Map