Assay Details
| Gene Symbol: AT5G24070 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122312.2 | NP_197795.1 | 1 - 2 | 406 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT5G24070| Gene Symbol: AT5G24070 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122312.2 | NP_197795.1 | 1 - 2 | 406 | 76 | |
Set Membership: |
|