Assay Details
Gene Symbol: SOS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203448.2 | NP_001190377.1 | 2 - 3 | 458 | 67 | |
NM_001343861.1 | NP_001331304.1 | 1 - 2 | 3275 | 67 | ||
NM_001343862.1 | NP_001331303.1 | 1 - 2 | 3275 | 67 | ||
NM_122333.6 | NP_197815.1 | 2 - 3 | 526 | 67 | ||
GenBank mRNA | AF192886.1 | - | 1 - 2 | 97 | 67 | |
AY063993.1 | - | 2 - 3 | 187 | 67 | ||
AY096693.1 | - | 1 - 2 | 97 | 67 | ||
Y18870.1 | - | 210 | 67 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001343861.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
