Assay Details
| Gene Symbol: SOS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203448.2 | NP_001190377.1 | 4 - 5 | 590 | 72 | |
| NM_001343861.1 | NP_001331304.1 | 3 - 4 | 3420 | 72 | ||
| NM_001343862.1 | NP_001331303.1 | 3 - 4 | 3407 | 72 | ||
| NM_001343863.1 | NP_001331302.1 | 2 - 3 | 873 | 72 | ||
| NM_122333.6 | NP_197815.1 | 4 - 5 | 658 | 72 | ||
| GenBank mRNA | AF192886.1 | - | 3 - 4 | 229 | 72 | |
| AY063993.1 | - | 4 - 5 | 319 | 72 | ||
| AY096693.1 | - | 3 - 4 | 229 | 72 | ||
| Y18870.1 | - | 342 | 72 | |||
More Information
Set Membership: |
|

SOS3
Genomic Map