Assay Details
| Gene Symbol: SS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203449.1 | NP_001190378.1 | 11 - 12 | 1740 | 75 | |
| NM_122336.5 | NP_197818.1 | 11 - 12 | 1964 | 75 | ||
| GenBank mRNA | AK226881.1 | - | 11 - 12 | 1673 | 75 | |
| AY128273.1 | - | 11 - 12 | 1662 | 75 | ||
| KJ138668.1 | - | 11 - 12 | 1596 | 75 | ||
More Information
Set Membership: |
|

SS1
Genomic Map