Assay Details
| Gene Symbol: SS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203449.1 | NP_001190378.1 | 4 - 5 | 1030 | 102 | |
| NM_122336.5 | NP_197818.1 | 4 - 5 | 1254 | 102 | ||
| GenBank mRNA | AK226881.1 | - | 4 - 5 | 963 | 102 | |
| AY128273.1 | - | 4 - 5 | 952 | 102 | ||
| KJ138668.1 | - | 4 - 5 | 886 | 102 | ||
More Information
Set Membership: |
|

SS1
Genomic Map