Assay Details
| Gene Symbol: AT5G24350 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203451.1 | NP_001190380.1 | 2 - 3 | 330 | 85 | |
| NM_001343876.1 | NP_001318636.1 | 2 - 3 | 345 | 85 | ||
| NM_122343.3 | NP_197823.1 | 2 - 3 | 330 | 85 | ||
More Information
Set Membership: |
|

AT5G24350
Genomic Map