Assay Details
| Gene Symbol: DMR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122361.4 | NP_197841.1 | 3 - 4 | 976 | 87 | |
| GenBank mRNA | AF386975.1 | - | 3 - 4 | 842 | 87 | |
| AY081455.1 | - | 3 - 4 | 771 | 87 | ||
More Information
Set Membership: |
|

DMR6
Genomic Map