Assay Details
| Gene Symbol: BIN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161263.1 | NP_001154735.1 | 4 - 5 | 646 | 106 | |
| NM_001161264.1 | NP_001154736.1 | 4 - 5 | 725 | 106 | ||
| NM_001161265.1 | NP_001154737.1 | 4 - 5 | 713 | 106 | ||
| NM_001161266.1 | NP_001154738.1 | 4 - 5 | 713 | 106 | ||
| NM_001161267.1 | NP_001154739.1 | 4 - 5 | 737 | 106 | ||
| NM_001343885.1 | NP_001330963.1 | 4 - 5 | 713 | 106 | ||
| NM_001343886.1 | NP_001330964.1 | 4 - 5 | 736 | 106 | ||
| NM_001343887.1 | NP_001330968.1 | 4 - 5 | 725 | 106 | ||
| NM_001343888.1 | NP_001330967.1 | 3 - 4 | 842 | 106 | ||
| NM_001343889.1 | NP_001330966.1 | 3 - 4 | 842 | 106 | ||
| NM_001343890.1 | NP_001330965.1 | 4 - 5 | 713 | 106 | ||
More Information
Set Membership: |
|

BIN4
Genomic Map