Assay Details
| Gene Symbol: AT5G24650 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122373.3 | NP_197853.1 | 5 - 6 | 837 | 64 | |
| GenBank mRNA | AY070735.1 | - | 5 - 6 | 776 | 64 | |
| AY142048.1 | - | 5 - 6 | 738 | 64 | ||
| DQ405267.1 | - | 5 - 6 | 738 | 64 | ||
More Information
Set Membership: |
|

AT5G24650
Genomic Map