Assay Details
| Gene Symbol: EMB2820 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203457.2 | NP_001190386.1 | 9 - 10 | 1485 | 88 | |
| NM_001343891.1 | NP_001318639.1 | 10 - 11 | 1853 | 88 | ||
| NM_001343892.1 | NP_001318640.1 | 9 - 10 | 1485 | 88 | ||
| NM_001343893.1 | NP_001330739.1 | 8 - 9 | 1547 | 88 | ||
| NM_122376.4 | NP_197855.3 | 9 - 10 | 1930 | 88 | ||
More Information
Set Membership: |
|

EMB2820
Genomic Map