Assay Details
| Gene Symbol: AT5G24750 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122384.3 | NP_568452.2 | 12 - 13 | 1368 | 55 | |
| GenBank mRNA | KJ676981.1 | - | 12 - 13 | 1225 | 55 | |
More Information
Set Membership: |
|

AT5G24750
Genomic Map