Assay Details
| Gene Symbol: AT5G24750 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122384.3 | NP_568452.2 | 1 - 2 | 231 | 65 | |
| GenBank mRNA | KJ676981.1 | - | 1 - 2 | 88 | 65 | |
More Information
Set Membership: |
|

AT5G24750
Genomic Map